Tisk…

α-Bromo-3-fluorotoulene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: α-Bromo-3-fluorotoluene, α-Bromo-m-fluorotoulene, alpha-Bromo-m-fluorotoulene, 1-(Bromomethyl)-3-fluorobenzene, m-Fluorobenzyl bromide, 3-Fluorobenzyl bromide

M-Clarity™ quality level = MQ100

F7504-25G F7504-5G
SIALF7504-25GEA 0 CZK
SIALF7504-25G SIALF7504-5G
α-Bromo-3-fluorotoulene, Sigma-Aldrich®
α-Bromo-3-fluorotoulene
Vzorec: C₇H₆BrF
MW: 189,03 g/mol
Bod varu: 88 °C (1013 hPa)
Hustota: 1,541 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000340
CAS číslo: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR