3,4,5-Trifluorobenzyl bromide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B22578.03
B22578.06
B22578.03EA
2030
CZK
B22578.03
B22578.06
3,4,5-Trifluorobenzyl bromide ≥97%
3,4,5-Trifluorobenzyl bromide
Purity: 97%
CAS No.: 220141-72-0
Molecular Formula: C7H4BrF3
Formula Weight: 225.01
Flash Point: 65°(149°F)
Refractive Index: 1.5050
Boiling Point: 44-46°/1mm
Beilstein Reference: 8680985
UN No.: 3265
MDL No.: MFCD00083528
Vzorec:
C₇H₄BrF₃ Bod varu: 44…46 °C (1 mmHg) Hustota: 1,689 Bod vzplanutí: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00083528 CAS číslo: 220141-72-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...