α-Bromo-2,5-difluorotoluene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide
M-Clarity™ quality level = MQ100
264423-10G
264423-1G
SIAL264423-10GEA
4660
CZK
SIAL264423-10G
SIAL264423-1G
α-Bromo-2,5-difluorotoluene, Sigma-Aldrich®
α-Bromo-2,5-difluorotoluene
Vzorec:
C₇H₅BrF₂ MW: 207,02 g/mol Bod varu: 186 °C (1013 hPa) Bod tání: 8 °C Hustota: 1,609 g/cm³ (20 °C) |
MDL Number:
MFCD00009897 CAS číslo: 85117-99-3 EINECS: 285-651-0 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...