Tisk…

2,4-Difluorobenzyl bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene

M-Clarity™ quality level = MQ200

264415-25G 264415-5G
SIAL264415-25GEA 8530 CZK
SIAL264415-25G SIAL264415-5G
2,4-Difluorobenzyl bromide, Sigma-Aldrich®
2,4-Difluorobenzyl bromide
Vzorec: C₇H₅BrF₂
MW: 207,02 g/mol
Bod varu: 42 °C (1 mmHg)
Hustota: 1,61 g/cm³ (20 °C)
MDL Number: MFCD00011649
CAS číslo: 23915-07-3
EINECS: 245-938-3
UN: 2920
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR