Tisk…

Pentafluorobenzyl bromide, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

101052-5G 101052-1G 101052-25G
SIAL101052-5GEA 2420 CZK
SIAL101052-5G SIAL101052-1G SIAL101052-25G
Pentafluorobenzyl bromide, Sigma-Aldrich®
Pentafluorobenzyl bromide
Vzorec: (C₆F₅)CH₂Br
Bod varu: 174 °C (1013 hPa)
Hustota: 1,72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS číslo: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR