Tisk…

3'-Bromoacetophenone ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: m-Bromoacetophenone

A12454.14 A12454.22 A12454.36
A12454.14EA 1360 CZK
A12454.14 A12454.22 A12454.36
3'-Bromoacetophenone ≥98%
3'-Bromoacetophenone

Purity: 98+% CAS No.: 2142-63-4 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 114°(237°F) Refractive Index: 1.5750 Melting Point: 7-8° Boiling Point: 79-81°/2mm Beilstein Reference: 507599 MDL No.: MFCD00000083

Vzorec: C₈H₇BrO
MW: 199,05 g/mol
Bod varu: 79…81 °C (2 mmHg)
Bod tání: 7…8 °C
Hustota: 1,498
Bod vzplanutí: 114 °C (237 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000083
CAS číslo: 2142-63-4
EINECS: 218-396-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR