(3-Bromophenyl)acetonitrile, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Bromobenzyl cyanide
M-Clarity™ quality level = MQ100
260088-25G
260088-5G
SIAL260088-25GEA
6040
CZK
SIAL260088-25G
SIAL260088-5G
(3-Bromophenyl)acetonitrile, Sigma-Aldrich®
(3-Bromophenyl)acetonitrile
Vzorec:
C₈H₆BrN MW: 196,05 g/mol Bod varu: 286 °C (1013 hPa) Bod tání: 27…28 °C |
MDL Number:
MFCD00001906 CAS číslo: 31938-07-5 EINECS: 250-867-6 UN: 3449 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...