(3-Bromophenyl)acetonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Bromobenzyl cyanide
B20189.06
B20189.14
B20189.22
B20189.06EA
1880
CZK
B20189.06
B20189.14
B20189.22
(3-Bromophenyl)acetonitrile ≥97%
(3-Bromophenyl)acetonitrile
Purity: 97%
CAS No.: 31938-07-5
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Flash Point: 126°(259°F)
Refractive Index: 1.5700
Melting Point: 25-28°
Boiling Point: 275°
Beilstein Reference: 2355439
UN No.: 3449
MDL No.: MFCD00001906
Vzorec:
C₈H₆BrN MW: 196,05 g/mol Bod varu: 275 °C Bod tání: 25…28 °C Bod vzplanutí: 126 °C (259 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001906 CAS číslo: 31938-07-5 EINECS: 250-867-6 UN: 3449 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...