(2-Bromophenyl)acetonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromobenzyl cyanide
A17548.06
A17548.14
A17548.06EA
1310
CZK
A17548.06
A17548.14
(2-Bromophenyl)acetonitrile ≥97%
(2-Bromophenyl)acetonitrile
Purity: 97%
CAS No.: 19472-74-3
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Flash Point: 113°(235°F)
Refractive Index: 1.5696
Odour: Pungent
Melting Point: 19°
Boiling Point: 252°
Beilstein Reference: 2043684
UN No.: 1694
MDL No.: MFCD00001896
Vzorec:
C₈H₆BrN MW: 196,05 g/mol Bod varu: 252 °C Bod tání: 19 °C Hustota: 1,510 Bod vzplanutí: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001896 CAS číslo: 19472-74-3 EINECS: 243-091-4 UN: 1694 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...