Tisk…

ω-Bromo-4-chloroacetophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Bromo-4'-chloroacetophenone, 2-Bromo-1-(4-chlorophenyl)ethanone, Bromomethyl p-chlorophenyl ketone, α-Bromo-4-chloroacetophenone, 2-Bromo-4′-chloroacetophenone, 4-Chlorophenacyl bromide

M-Clarity™ quality level = MQ100

101273-100G 101273-25G
SIAL101273-100GEA 4160 CZK
SIAL101273-100G SIAL101273-25G
ω-Bromo-4-chloroacetophenone, Sigma-Aldrich®
ω-Bromo-4-chloroacetophenone
Vzorec: C₈H₆BrClO
MW: 233.49
Bod varu: 278
Bod tání: 110
MDL Number: MFCD00000625
CAS číslo: 536-38-9
EINECS: 208-631-5
UN: 3261
ADR: 8 III
Merck Index: 13,02172

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR