Tisk…

ω-Bromo-2-chloroacetophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

594482-1G 594482-5G
SIAL594482-1GEA 3970 CZK
SIAL594482-1G SIAL594482-5G
ω-Bromo-2-chloroacetophenone, Sigma-Aldrich®
ω-Bromo-2-chloroacetophenone
Vzorec: C₈H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD00832993
CAS číslo: 5000-66-8
UN: 1760
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR