Tisk…

(3-Formylfenyl)boronová kyselina

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Dihydroxy(3-formylphenyl)borane, 3-Boronobenzaldehyde, (3-Formylphenyl)metaboric acid, 3-Formylbenzeneboronic acid, (3-Formylphenyl)boronic acid

B25437.03 B25437.06 B25437.14
B25437.03EA 1390 CZK
B25437.03 B25437.06 B25437.14
(3-Formylfenyl)boronová kyselina
(3-Formylfenyl)boronová kyselina

CAS No.: 87199-16-4 Molecular Formula: C7H7BO3 Formula Weight: 149.94 Melting Point: 109-113° Beilstein Reference: 3030769 MDL No.: MFCD00161356

Vzorec: C₇H₇BO₃
MW: 149.94
Bod tání: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00161356
CAS číslo: 87199-16-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR