(2-Formylfenyl)boronová kyselina, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane
M-Clarity™ quality level = MQ100
431958-1G
431958-5G
SIAL431958-1GEA
1300
CZK
SIAL431958-1G
SIAL431958-5G
(2-Formylfenyl)boronová kyselina, Sigma-Aldrich®
(2-Formylfenyl)boronová kyselina
Vzorec:
C₇H₇BO₃ MW: 149.94 |
MDL Number:
MFCD00151822 CAS číslo: 40138-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...