Diphenylacetylene 99%
Dodavatel: Thermo Scientific
117180050
117180250
117181000
ACRO117180050EA
1200
CZK
ACRO117180050
ACRO117180250
ACRO117181000
Diphenylacetylene 99%
Diphenylacetylene
Vzorec:
C₁₄H₁₀ MW: 178,23 g/mol Bod varu: 170 °C (19 mmHg) Bod tání: 58…61 °C Hustota: 0,99 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00004786 CAS číslo: 501-65-5 EINECS: 207-926-6 Merck Index: 12,09643 |
Výsledky specifikačního testu
Appearance | Light yellow to brown Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 58 to 61 °C |
GC | ≥98.5 % |
Calcium (Ca) | ≤100 ppm |
Magnesium (Mg) | ≤50 ppm |
Potassium (K) | ≤50 ppm |
Sodium (Na) | ≤500 ppm |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...