1-Phenyl-1-propyne, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
1-Propynylbenzene, Prop-1-yn-1-ylbenzene
M-Clarity™ quality level = MQ100
161241-25G
161241-5G
SIAL161241-25GEA
6870
CZK
SIAL161241-25G
SIAL161241-5G
1-Phenyl-1-propyne, Sigma-Aldrich®
1-Phenyl-1-propyne
Vzorec:
C₆H₅CCCH₃ MW: 116,16 g/mol Bod varu: 185 °C (1013 hPa) Hustota: 0,942 g/cm³ (15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009272 CAS číslo: 673-32-5 EINECS: 211-607-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...