4-Ethynylanisole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(4-Methoxyphenyl)ethyne, (4-Methoxyphenyl)acetylene, 1-Ethynyl-4-methoxybenzene, 1-Methoxy-4-ethynylbenzene
206490-1G
206490-5G
SIAL206490-1GEA
2830
CZK
SIAL206490-1G
SIAL206490-5G
4-Ethynylanisole, Sigma-Aldrich®
4-Ethynylanisole
Vzorec:
C₉H₈O MW: 132,16 g/mol |
MDL Number:
MFCD00168815 CAS číslo: 768-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...