1,1-Diphenylethylene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
M-Clarity™ quality level = MQ100
D206806-10G
D206806-25G
D206806-5G
SIALD206806-10GEA
1970
CZK
SIALD206806-10G
SIALD206806-25G
SIALD206806-5G
1,1-Diphenylethylene, Sigma-Aldrich®
1,1-Diphenylethylene
Vzorec:
(C₆H₅)₂CCH₂ MW: 180.25 Bod varu: 270 Hustota: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008583 CAS číslo: 530-48-3 EINECS: 208-482-6 Merck Index: 13,03355 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...