4-Allyl-2,6-dimethoxyphenol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W365505-100G
W365505-1KG
W365505-25G
W365505-SAMPLE
SIALW365505-100GEA
35020
CZK
SIALW365505-100G
SIALW365505-1KG
SIALW365505-25G
SIALW365505-SAMPLE
4-Allyl-2,6-dimethoxyphenol, Sigma-Aldrich®
4-Allyl-2,6-dimethoxyphenol
Vzorec:
C₁₁H₁₄O₃ MW: 194,23 g/mol |
MDL Number:
MFCD00008655 CAS číslo: 6627-88-9 EINECS: 229-600-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...