Tisk…

1,2-Benzenedisulphonyl chloride

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Benzene-1,2-di(sulphonyl chloride)

H50375.03 H50375.06
H50375.03EA 4190 CZK
H50375.03 H50375.06
1,2-Benzenedisulphonyl chloride
1,2-Benzenedisulphonyl chloride

CAS No.: 6461-76-3 Molecular Formula: C6H4Cl2O4S2 Formula Weight: 275.13 Melting Point: 140-146° UN No.: 3261 MDL No.: MFCD00059836

Vzorec: C₆H₄Cl₂O₄S₂
Bod tání: 140…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00059836
CAS číslo: 6461-76-3
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR