Tisk…

Thioanisole, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene

T28002-100G T28002-25G T28002-500G
SIALT28002-100GEA 1790 CZK
SIALT28002-100G SIALT28002-25G SIALT28002-500G
Thioanisole, Sigma-Aldrich®
Thioanisole
Vzorec: C₇H₈S
MW: 124,21 g/mol
Bod varu: 188 °C (1013 hPa)
Bod tání: –15 °C
Hustota: 1,05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008559
CAS číslo: 100-68-5
EINECS: 202-878-2
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR