Thioanisole ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene
A14846.0E
A14846.14
A14846.22
A14846.36
A14846.0EEA
20080
CZK
A14846.0E
A14846.14
A14846.22
A14846.36
Thioanisole ≥99%
Thioanisole
Purity: 99%
CAS No.: 100-68-5
Molecular Formula: C7H8S
Formula Weight: 124.21
Flash Point: 57°(135°F)
Refractive Index: 1.5860
Odour: Stench
Melting Point: -15°
Boiling Point: 188-189°
Beilstein Reference: 1904179
UN No.: 1993
MDL No.: MFCD00008559
Vzorec:
C₇H₈S MW: 124,21 g/mol Bod varu: 188…189 °C Bod tání: –15 °C Hustota: 1,058 Bod vzplanutí: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008559 CAS číslo: 100-68-5 EINECS: 202-878-2 UN: 3334 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...