Tisk…

Difenylether, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diphenyl oxide, Phenyl ether, Phenoxybenzene

M-Clarity™ quality level = MQ100

240834-100G 240834-5G
SIAL240834-100GEA 1930 CZK
SIAL240834-100G SIAL240834-5G
Difenylether, Sigma-Aldrich®
Difenylether
Vzorec: C₁₂H₁₀O
MW: 170,21 g/mol
Bod varu: 498 °C (1013 hPa)
Bod tání: 24…27 °C
Hustota: 1,07 g/cm³ (20 °C)
Bod vzplanutí: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS číslo: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: 9,III
Merck Index: 12,07442

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR