4-Hydroxythiophenol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L04429.06
L04429.14
L04429.06EA
2360
CZK
L04429.06
L04429.14
4-Hydroxythiophenol ≥97%
4-Hydroxythiophenol
Purity: 97%
CAS No.: 637-89-8
Molecular Formula: C6H6OS
Formula Weight: 126.18
Flash Point: 110°(230°F)
Odour: Unpleasant
Boiling Point: 256°
Beilstein Reference: 2039306
UN No.: 1759
MDL No.: MFCD00004850
Vzorec:
C₆H₆OS Bod varu: 256 °C Bod tání: 33…37 °C Hustota: 1,255 Bod vzplanutí: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004850 CAS číslo: 637-89-8 EINECS: 211-307-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...