Tisk…

Fenol, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

M-Clarity™ quality level = MQ200

185450-100G 185450-2.5KG 185450-500G
SIAL185450-100GEA 707 CZK
SIAL185450-100G SIAL185450-2.5KG SIAL185450-500G
Fenol, Sigma-Aldrich®
Fenol
Vzorec: C₆H₅OH
MW: 94.11
Bod varu: 181.7
Bod tání: 40.8
Hustota: 1.0722 g/cm³ (20 °C)
Bod vzplanutí: 81
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS číslo: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
REACH: 01-2119471329-32
Merck Index: 12,07390

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR