1,3,5-Trimethoxybenzene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
O,O,O-1,3,5-Trimethylresorcinol
M-Clarity™ quality level = MQ200
138827-10G
138827-250G
138827-50G
SIAL138827-10GEA
928
CZK
SIAL138827-10G
SIAL138827-250G
SIAL138827-50G
1,3,5-Trimethoxybenzene, Sigma-Aldrich®
1,3,5-Trimethoxybenzene
Vzorec:
C₉H₁₂O₃ MW: 168,19 g/mol Bod varu: 255 °C (1013 hPa) Bod tání: 52…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008385 CAS číslo: 621-23-8 EINECS: 210-673-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...