2,4-Dinitrobenzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,4-Dinitrobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ200
142956-25G
142956-5G
SIAL142956-25GEA
0
CZK
SIAL142956-25G
SIAL142956-5G
2,4-Dinitrobenzenesulphonyl chloride, Sigma-Aldrich®
2,4-Dinitrobenzenesulphonyl chloride
Vzorec:
C₆H₃ClN₂O₆S MW: 266,62 g/mol Bod tání: 102 °C |
MDL Number:
MFCD00007431 CAS číslo: 1656-44-6 EINECS: 216-749-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...