Tisk…

4-Nitrothioanisole ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Nitrophenyl methyl sulphide, 1-Nitro-4-(methylthio)benzene

B20339.06 B20339.14
B20339.06EA 1300 CZK
B20339.06 B20339.14
4-Nitrothioanisole ≥98%
4-Nitrothioanisole

Purity: 98% CAS No.: 701-57-5 Molecular Formula: C7H7NO2S Formula Weight: 169.20 Melting Point: 66-72° Beilstein Reference: 1817928

Vzorec: C₇H₇NO₂S
MW: 169,2 g/mol
Bod varu: 135…140 °C (2 mmHg)
Bod tání: 66…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00010868
CAS číslo: 701-57-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR