Tisk…

Difenylamin ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N-Phenylaniline, N-Phenylbenzeneamine

A18265.0E A18265.22 A18265.36
A18265.0EEA 5430 CZK
A18265.0E A18265.22 A18265.36
Difenylamin ≥98%
Difenylamin

Purity: 98+% CAS No.: 122-39-4 Molecular Formula: C12H11N Formula Weight: 169.23 Flash Point: 152°(305°F) Odour: Weak Boiling Point: 302° Beilstein Reference: 508755 Merck Reference: 14,3317 UN No.: 3077 MDL No.: MFCD00003014

Vzorec: (C₆H₅)₂NH
MW: 169,23 g/mol
Bod varu: 302 °C
Bod tání: 52…55 °C
Hustota: 1,16
Bod vzplanutí: 152 °C (305 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS číslo: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
Merck Index: 14,3317
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR