Difenylamin ACS
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
N-Phenylaniline, N-Phenylbenzeneamine
36610.14
36610.22
36610.36
36610.14EA
1270
CZK
36610.14
36610.22
36610.36
Difenylamin ACS
Difenylamin
CAS No.: 122-39-4
Molecular Formula: C12H11N
Formula Weight: 169.23
Flash Point: 152°(305°F)
Physical Form: Solid
Density: 1.16
Appearance: White to off-white
Melting Point: 52-55°
Boiling Point: 302°
Solubility: Soluble in benzene, ether, glacial acetic acid, CS2
Beilstein Reference: 508755
Merck Reference: 14,3317
UN No.: 3077
MDL No.: MFCD00003014
Vzorec:
(C₆H₅)₂NH MW: 169,23 g/mol Bod varu: 302 °C Bod tání: 52…55 °C Hustota: 1,16 Bod vzplanutí: 152 °C (305 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003014 CAS číslo: 122-39-4 EINECS: 204-539-4 UN: 3077 ADR: 9,III Merck Index: 14,3317 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...