Tisk…

N-Methyl-4-nitroaniline ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15548.06 A15548.14 A15548.22
A15548.06EA 776 CZK
A15548.06 A15548.14 A15548.22
N-Methyl-4-nitroaniline ≥97%
N-Methyl-4-nitroaniline

Purity: 97% CAS No.: 100-15-2 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Odour: Characteristic Melting Point: 147-154° Beilstein Reference: 637920 UN No.: 2811 MDL No.: MFCD00007305

Vzorec: C₇H₈N₂O₂
MW: 152.15
Bod varu: 291
Bod tání: 147…154 °C
Hustota: 1.201 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007305
CAS číslo: 100-15-2
EINECS: 202-823-2
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR