4-Nitro-m-phenylenediamine 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Nitro-1,3-phenylenediamine
L12166.09
L12166.18
L12166.09EA
1460
CZK
L12166.09
L12166.18
4-Nitro-m-phenylenediamine 95%
4-Nitro-m-phenylenediamine
Purity: 95%
CAS No.: 5131-58-8
Molecular Formula: C6H7N3O2
Formula Weight: 153.14
Melting Point: 156-160°
Beilstein Reference: 2094904
Vzorec:
C₆H₇N₃O₂ MW: 153,14 g/mol Bod tání: 156…160 °C Hustota: 1,552 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00025289 CAS číslo: 5131-58-8 EINECS: 225-876-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...