m-Fenylendiamin 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
MPD, 1,3-Diaminobenzene, 1,3-Phenylenediamine
A15339.0B
A15339.30
A15339.0BEA
3240
CZK
A15339.0B
A15339.30
m-Fenylendiamin 98%
m-Fenylendiamin
Purity: 98%
CAS No.: 108-45-2
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 187°(368°F)
Melting Point: 62-66°
Boiling Point: 284-287°
Solubility: Soluble in water, methanol, acetone, DMF, MEK, dioxane
Beilstein Reference: 471357
Merck Reference: 14,7283
UN No.: 1673
MDL No.: MFCD00007799
Vzorec:
C₆H₈N₂ Bod varu: 284…287 °C Bod tání: 62…66 °C Hustota: 1,139 Bod vzplanutí: 187 °C (368 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007799 CAS číslo: 108-45-2 EINECS: 203-584-7 UN: 1673 ADR: 6.1,III Merck Index: 14,7283 |