Tisk…

Kyselina sulfanilová ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A15825.30 A15825.0B A15825.0I
A15825.30EA 1620 CZK
A15825.30 A15825.0B A15825.0I
Kyselina sulfanilová ≥98%
Kyselina sulfanilová

Purity: 98+% CAS No.: 121-57-3 Molecular Formula: C6H7NO3S Formula Weight: 173.19 Physical Form: Cryst powder Density: 1.485 Appearance: White to pale brown Melting Point: ca 365° Beilstein Reference: 908765 Merck Reference: 14,8926 MDL No.: MFCD00007886


Reagent for the detection of histidine; intermediate for dyes and pharmaceuticals

Vzorec: 4-(H₂N)C₆H₄SO₃H
Bod varu: 363 °C (1013 hPa)
Bod tání: ∼365 °C
Hustota: 1,485
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS číslo: 121-57-3
EINECS: 204-482-5
Merck Index: 14,8926
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR