Tisk…

Sulfanilamid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Aminobenzenesulphonamide

M-Clarity™ quality level = MQ100

33626-100G 33626-6X100G
SIAL33626-100GEA 4350 CZK
SIAL33626-100G SIAL33626-6X100G
Sulfanilamid, Sigma-Aldrich®
Sulfanilamid
Vzorec: H₂NC₆H₄SO₂NH₂
MW: 172,21 g/mol
Bod varu: 401 °C (1013 hPa)
Bod tání: 163…167 °C
Hustota: 1,46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS číslo: 63-74-1
EINECS: 200-563-4
Merck Index: 13,09010
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR