Sulfanilamid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A13001.0E
A13001.22
A13001.36
A13001.0EEA
10850
CZK
A13001.0E
A13001.22
A13001.36
Sulfanilamid ≥98%
Sulfanilamid
Purity: 98%
CAS No.: 63-74-1
Molecular Formula: C6H8N2O2S
Formula Weight: 172.21
Melting Point: 163-168°
Beilstein Reference: 511852
Merck Reference: 14,8925
MDL No.: MFCD00007939
Vzorec:
H₂NC₆H₄SO₂NH₂ Bod varu: 401 °C (1013 hPa) Bod tání: 163…168 °C Hustota: 1,08 Storage Temperature: Ambient |
MDL Number:
MFCD00007939 CAS číslo: 63-74-1 EINECS: 200-563-4 Merck Index: 14,8925 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...