Tisk…

3-Aminothiophenol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

143480-25G 143480-5G
SIAL143480-25GEA 9750 CZK
SIAL143480-25G SIAL143480-5G
3-Aminothiophenol, Sigma-Aldrich®
3-Aminothiophenol
Vzorec: C₆H₇NS
Bod varu: 110…112 °C (1 mmHg)
Hustota: 1,1859 g/cm³ (20 °C)
MDL Number: MFCD00007791
CAS číslo: 22948-02-3
EINECS: 245-344-4
UN: 3267
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR