o-Anisidin ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Anisidine, 2-Aminoanisole, 2-Methoxyaniline
A10288.0B
A10288.0I
A10288.30
AB113747.1000
A10288.0BEA
3250
CZK
A10288.0B
A10288.0I
A10288.30
ABCRAB113747.1000
o-Anisidin ≥99%
o-Anisidin
Purity: 99%
CAS No.: 90-04-0
Molecular Formula: C7H9NO
Formula Weight: 123.16
Flash Point: 98°(208°F)
Physical Form: Liquid
Density: 1.092
Refractive Index: 1.5740
Sensitivity: Light Sensitive
Appearance: Yellow to brown
Melting Point: 5-6°
Boiling Point: 224-225°
Beilstein Reference: 386210
Merck Reference: 14,667
UN No.: 2431
MDL No.: MFCD00007688
Vzorec:
C₇H₉NO Bod varu: 224…225 °C Bod tání: 5…6 °C Hustota: 1,092 Bod vzplanutí: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007688 CAS číslo: 90-04-0 EINECS: 201-963-1 UN: 2431 ADR: 6.1,III Merck Index: 14,667 |