Tisk…

4-Nitrophenyl isothiocyanate ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

L11042.09 L11042.18 L11042.30
L11042.09EA 2050 CZK
L11042.09 L11042.18 L11042.30
4-Nitrophenyl isothiocyanate ≥97%
4-Nitrophenyl isothiocyanate

Purity: 97% CAS No.: 2131-61-5 Molecular Formula: C7H4N2O2S Formula Weight: 180.19 Odour: Irritating Melting Point: 105-110° Boiling Point: 137-138°/11mm Beilstein Reference: 640027 UN No.: 2923 MDL No.: MFCD00007307

Vzorec: C₇H₄N₂O₂S
MW: 180,19 g/mol
Bod varu: 137…138 °C (11 mmHg)
Bod tání: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007307
CAS číslo: 2131-61-5
EINECS: 218-359-9
UN: 2923
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR