Fenylizothiokyanát ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A11596.06
A11596.22
A11596.36
A11596.06EA
996
CZK
A11596.06
A11596.22
A11596.36
Fenylizothiokyanát ≥97%
Fenylizothiokyanát
Purity: 97%
CAS No.: 103-72-0
Molecular Formula: C7H5NS
Formula Weight: 135.19
Flash Point: 87°(188°F)
Refractive Index: 1.6500
Odour: Pungent
Melting Point: -21°
Boiling Point: 220-221°
Beilstein Reference: 471392
Merck Reference: 14,7297
UN No.: 2927
MDL No.: MFCD00004798
Vzorec:
C₇H₅NS Bod varu: 220…221 °C Bod tání: –21 °C Hustota: 1,131 Bod vzplanutí: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004798 CAS číslo: 103-72-0 EINECS: 203-138-1 UN: 2927 ADR: 6.1,II Merck Index: 14,7297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...