Tisk…

3-Methoxyphenyl isocyanate ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B24329.06 B24329.14
B24329.06EA 1820 CZK
B24329.06 B24329.14
3-Methoxyphenyl isocyanate ≥99%
3-Methoxyphenyl isocyanate

Purity: 99% CAS No.: 18908-07-1 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Flash Point: 96°(204°F) Refractive Index: 1.5430 Odour: Pungent Boiling Point: 94-95°/10mm Beilstein Reference: 775986 UN No.: 2206 MDL No.: MFCD00002019

Vzorec: C₈H₇NO₂
Bod varu: 94…95 °C (10 mmHg)
Hustota: 1,138
Bod vzplanutí: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002019
CAS číslo: 18908-07-1
EINECS: 242-658-3
UN: 2206
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR