4-Nitrophenyl isocyanate ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Isocyanato-4-nitrobenzene
A11194.03
A11194.06
A11194.14
A11194.03EA
913
CZK
A11194.03
A11194.06
A11194.14
4-Nitrophenyl isocyanate ≥97%
4-Nitrophenyl isocyanate
Purity: 97%
CAS No.: 100-28-7
Molecular Formula: C7H4N2O3
Formula Weight: 164.12
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₇H₄N₂O₃ MW: 164,12 g/mol Bod varu: 137…138 °C (11 mmHg) Bod tání: 56…59 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007306 CAS číslo: 100-28-7 EINECS: 202-836-3 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...