3-Nitrophenyl isocyanate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-Isocyanato-3-nitrobenzene, 3-Nitrophenylisocyanate
A16920.06
A16920.14
A16920.06EA
1730
CZK
A16920.06
A16920.14
3-Nitrophenyl isocyanate ≥97%
3-Nitrophenyl isocyanate
Purity: 97%
CAS No.: 3320-87-4
Molecular Formula: C7H4N2O3
Formula Weight: 164.12
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₇H₄N₂O₃ Bod varu: 130…131 °C (11 mmHg) Bod tání: 51…53 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007220 CAS číslo: 3320-87-4 EINECS: 222-025-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...