N,N,N',N'-Tetramethyl-p-phenylenediamine, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
N,N,N,N-Tetramethyl-1,4-phenylenediamine, N,N,N′,N′-Tetramethyl-p-phenylenediamine, TMPD, N,N,N',N'-Tetramethyl-1,4-phenylenediamine
T7394-25G
T7394-5G
SIALT7394-25GEA
5020
CZK
SIALT7394-25G
SIALT7394-5G
N,N,N',N'-Tetramethyl-p-phenylenediamine, Sigma-Aldrich®
N,N,N',N'-Tetramethyl-p-phenylenediamine
Vzorec:
C₁₀H₁₆N₂ Bod varu: 260 °C (1013 hPa) Bod tání: 48…50 °C Hustota: 1,08 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008309 CAS číslo: 100-22-1 EINECS: 202-831-6 UN: 1673 ADR: 6.1,III Merck Index: 12,09367 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...