Tisk…

3-(Diethylamino)phenol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Diethylaminophenol, N,N-Diethyl-3-aminophenol, N,N-Diethyl-3-hydroxyaniline

M-Clarity™ quality level = MQ100

102091-100G 102091-500G
SIAL102091-100GEA 1310 CZK
SIAL102091-100G SIAL102091-500G
3-(Diethylamino)phenol, Sigma-Aldrich®
3-(Diethylamino)phenol
Vzorec: C₁₀H₁₅NO
MW: 165,24 g/mol
Bod varu: 276 °C (1013 hPa)
Bod tání: 78 °C
MDL Number: MFCD00002265
CAS číslo: 91-68-9
EINECS: 202-090-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR