Tisk…

4-Amino-N,N-dimethylaniline 96%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N,N-Dimethylbenzene-p-diamine, N,N-Dimethyl-1,4-phenylenediamine, N,N-Dimethylbenzene-1,4-diamine, N,N-Dimethyl-p-phenylenediamine

A15962.0B A15962.18 A15962.30
A15962.0BEA 19630 CZK
A15962.0B A15962.18 A15962.30
4-Amino-N,N-dimethylaniline 96%
4-Amino-N,N-dimethylaniline

Purity: 96% CAS No.: 99-98-9 Molecular Formula: C8H12N2 Formula Weight: 136.20 Flash Point: 130°(266°F) Melting Point: 34-38° Boiling Point: 262° Beilstein Reference: 508105 Merck Reference: 14,3254 UN No.: 2811 MDL No.: MFCD00007860

Vzorec: C₈H₁₂N₂
MW: 136,2 g/mol
Bod varu: 262 °C
Bod tání: 34…38 °C
Hustota: 1,09
Bod vzplanutí: 130 °C (266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007860
CAS číslo: 99-98-9
EINECS: 202-807-5
UN: 2811
ADR: 6.1,II
Merck Index: 14,3254

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR