N,N-Dimethylanilin, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
407275-100ML
407275-800ML
SIAL407275-100MLEA
2980
CZK
SIAL407275-100ML
SIAL407275-800ML
N,N-Dimethylanilin, Sigma-Aldrich®
N,N-Dimethylanilin
Vzorec:
C₈H₁₁N MW: 121.18 Bod varu: 194 Bod tání: 2.5 Hustota: 0.9559 g/cm³ (20 °C) Bod vzplanutí: 62 Storage Temperature: Ambient |
MDL Number:
MFCD00008304 CAS číslo: 121-69-7 EINECS: 204-493-5 UN: 2253 ADR: 6.1 II Merck Index: 13,03261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...