Tisk…

Azobenzen ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A10425.14 A10425.22 A10425.36
A10425.14EA 1830 CZK
A10425.14 A10425.22 A10425.36
Azobenzen ≥97%
Azobenzene

Purity: 97+% CAS No.: 103-33-3 Molecular Formula: C12H10N2 Formula Weight: 182.23 Melting Point: 65-69° Boiling Point: 293° Merck Reference: 14,917 UN No.: 3077 MDL No.: MFCD00003022

Vzorec: C₁₂H₁₀N₂
Bod varu: 293 °C
Bod tání: 65…69 °C
Hustota: 1,203
Bod vzplanutí: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003022
CAS číslo: 103-33-3
EINECS: 203-102-5
UN: 3077
ADR: 9,III
Merck Index: 14,917

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR