3-Iodoanisole, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
3-iodoanisole
M-Clarity™ quality level = MQ100
242691-100G
242691-25G
242691-5G
SIAL242691-100GEA
11200
CZK
SIAL242691-100G
SIAL242691-25G
SIAL242691-5G
3-Iodoanisole, Sigma-Aldrich®
3-Iodoanisole
Vzorec:
C₇H₇IO MW: 234.04 Bod varu: 245 Hustota: 1.765 |
MDL Number:
MFCD00001048 CAS číslo: 766-85-8 EINECS: 212-173-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...