2-Iodoanisole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
252786-100G
252786-25G
SIAL252786-100GEA
3920
CZK
SIAL252786-100G
SIAL252786-25G
2-Iodoanisole, Sigma-Aldrich®
2-Iodoanisole
Vzorec:
C₇H₇IO Bod varu: 238 °C (1013 hPa) Hustota: 1,445 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001039 CAS číslo: 529-28-2 EINECS: 208-456-4 Merck Index: 13,05050 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...