Tisk…

4-Iodo-3-nitroanisole ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Iodo-4-methoxy-2-nitrobenzene

L11714.06 L11714.14
L11714.06EA 2340 CZK
L11714.06 L11714.14
4-Iodo-3-nitroanisole ≥98%
4-Iodo-3-nitroanisole

Purity: 98+% CAS No.: 58755-70-7 Molecular Formula: C7H6INO3 Formula Weight: 279.04 Melting Point: 60-63° Beilstein Reference: 2365592

Vzorec: C₇H₆INO₃
Bod tání: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00024195
CAS číslo: 58755-70-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR